Anti-SPRR3 Rabbit Polyclonal Antibody [clone: ]
Katalognr. ADIPAG25A0116C100
Anbieter: AdipoGen Life Sciences
Technische Daten
- Nom de l'antigène:Small Proline-Rich Protein 3
- Symbole de l'antigène:SPRR3
- Clonalité:Polyclonal
- Hôte:Rabbit
- ImmunoChimie:Yes
- Réactivité:Human
- Western blot:Yes
- Formulaire:Liquid
- Synonymes antigène:22 kDa pancornulin|Small proline rich protein 3|Small proline-rich protein 3|SPRC|Cornin beta|Cornin B|Sprr3|SPRR3_HUMAN.|Esophagin
- Storage buffer:Neat serum containing 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μL
- Cdt:100 µG
Technische Daten
Über diesen Artikel
Type:
Antigen: SPRR3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human