Anti-MDA5 Rabbit Polyclonal Antibody [clone: ]
Référence ADIPAG25A0114C100
Fournisseur: AdipoGen Life Sciences
Spécifications
- Symbole de l'antigène:MDA5
- Clonalité:Polyclonal
- Hôte:Rabbit
- ImmunoChimie:Yes
- Réactivité:Mouse
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:Neat serum containing 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μL
- Cdt:100 µG
Spécifications
À propos de cet article
Type:
Antigen: MDA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse