Anti-CTRP3 Guinea Pig Polyclonal Antibody [clone: ]
N° de catalogue ADIPAG25A0103C100
Fournisseur: AdipoGen Life Sciences
Spécifications
- Nom de l'antigène:C1Q And Tumor Necrosis Factor Related Protein 3
- Symbole de l'antigène:CTRP3
- Clonalité:Polyclonal
- ELISA:Yes
- Hôte:Guinea pig
- Réactivité:Human
- Western blot:Yes
- Formulaire:Liquid
- Synonymes antigène:CORS|CTRP3|C1ATNF3|CORS26|CORCS|CORS-26
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Spécifications
À propos de cet article
Type:
Antigen: CTRP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Human