Anti-GRN Rat Polyclonal Antibody [clone: ]
N° de catalogue ADIPAG25A0089C100
Fournisseur: AdipoGen Life Sciences
Spécifications
- Nom de l'antigène:Progranulin
- Symbole de l'antigène:GRN
- Clonalité:Polyclonal
- ELISA:Yes
- Hôte:Rat
- Réactivité:Mouse
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:0,7 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Spécifications
À propos de cet article
Type:
Antigen: GRN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse