Anti-SENP2 Rabbit Polyclonal Antibody [clone: ]
Catalog # ADIPAG25A0074C100
Supplier: AdipoGen Life Sciences
Specifications
- Nom de l'antigène:Sentrin-specific protease 2
- Symbole de l'antigène:SENP2
- Clonalité:Polyclonal
- ELISA:Yes
- Hôte:Rabbit
- Réactivité:Mouse
- Western blot:Yes
- Formulaire:Liquid
- Synonymes antigène:FKHL6|SUMO1/Sentrin/SMT3 Specific Peptidase 2|FREAC2|FREAC-2
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Specifications
About this item
Type:
Antigen: SENP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse