Anti-RBP4 Guinea Pig Polyclonal Antibody [clone: ]
ADIPAG25A0049C100
- Nom de l'antigène:Retinol binding protein 4
- Symbole de l'antigène:RBP4
- Clonalité:Polyclonal
- ELISA:Yes
- Hôte:Guinea pig
- Réactivité:Human
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Type:
Antigen: RBP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Human