Anti-GITRL Rat Polyclonal Antibody [clone: ]
ADIPAG25A0020C100
- Nom de l'antigène:Tumor Necrosis Factor (Ligand) Superfamily, Member 18
- Symbole de l'antigène:GITRL
- Clonalité:Polyclonal
- ELISA:Yes
- Hôte:Rat
- Réactivité:Mouse
- Western blot:Yes
- Formulaire:Liquid
- Synonymes antigène:hGITRL|TL6|AITRL|GITRL
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Type:
Antigen: GITRL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse