Anti-GITR Rat Polyclonal Antibody [clone: ]
Catalog # ADIPAG25A0014C100
Anbieter: AdipoGen Life Sciences
Technische Daten
- Nom de l'antigène:Tumor necrosis Factor Receptor superfamily member 18
- Symbole de l'antigène:GITR
- Clonalité:Polyclonal
- ELISA:Yes
- Hôte:Rat
- Réactivité:Human
- Formulaire:Liquid
- Synonymes antigène:CD357|Tumor necrosis factor receptor superfamily member 18|GITR|Glucocorticoid-induced TNFR-related protein|TNFRSF18|Activation-inducible TNFR family receptor|UNQ319|AITR|PRO364
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Technische Daten
Über diesen Artikel
Type:
Antigen: GITR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human