Anti-TRAIL-R4 Mouse Monoclonal Antibody [clone: HS402]
ADIPAG20B0023BC100
- Symbole de l'antigène:TRAIL-R4
- Clonalité:Monoclonal
- Clone:HS402
- Hôte:Mouse
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Isotype:IgG1
- Réactivité:Human
- Formulaire:Liquid
- Storage buffer:In PBS containing 10% glycerol and 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Type:
Antigen: TRAIL-R4
Clonality: Monoclonal
Clone: HS402
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human