Anti-TRAIL-R3 Mouse Monoclonal Antibody [clone: HS301]
ADIPAG20B0022FT100
- Nom de l'antigène:TNF receptor superfamily member 10C
- Symbole de l'antigène:TNFRSF10C
- Clonalité:Monoclonal
- Clone:HS301
- Hôte:Mouse
- ImmunoChimie:Yes
- Isotype:IgG1
- Réactivité:Human
- Formulaire:Liquid
- Synonymes antigène:DCR1|TRID|TRAIL-R3|TRAILR3|DCR1-TNFR|CD263|LIT
- Storage buffer:In PBS containing 10% glycerol and 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 T
Type:
Antigen: TRAIL-R3
Clonality: Monoclonal
Clone: HS301
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human