Anti-TRAIL-R2 Mouse Monoclonal Antibody [clone: HS201]
ADIPAG20B0022BC100
- Nom de l'antigène:TNF receptor superfamily member 10B
- Symbole de l'antigène:TNFRSF10B
- Clonalité:Monoclonal
- Clone:HS201
- Hôte:Mouse
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Isotype:IgG1
- Réactivité:Human
- Formulaire:Liquid
- Synonymes antigène:TRAILR2|TRAIL-R2|CD262|Trail receptor 2|TRICK2|KILLER|TRICK2A|TRICKB|ZTNFR9|KILLER/DR5|TRICK2B|DR5
- Storage buffer:In PBS containing 10% glycerol and 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Type:
Antigen: TRAIL-R2
Clonality: Monoclonal
Clone: HS201
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human