Anti-Polyglutamylation Mouse Monoclonal Antibody (Biotin) [clone: GT335]
ADIPAG20B0016C100
- Symbole de l'antigène:Polyglutamylation
- Clonalité:Monoclonal
- Clone:GT335
- Conjugaison:Biotin
- Hôte:Mouse
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Isotype:IgG1 kappa
- Réactivité:Multiple
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:In PBS containing 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Type:
Antigen: Polyglutamylation
Clonality: Monoclonal
Clone: GT335
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: All species