Anti-BAFF-R Mouse Monoclonal Antibody (Biotin) [clone: HuBR9.1]
Catalog # ADIPAG20B0012C100
Supplier: AdipoGen Life Sciences
Specifications
- Symbole de l'antigène:BAFF-R
- Clonalité:Monoclonal
- Clone:HuBR9.1
- Conjugaison:Biotin
- Hôte:Mouse
- Isotype:IgG1
- Réactivité:Human
- Formulaire:Liquid
- Storage buffer:In PBS containing 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:0,5 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:100 µG
Specifications
About this item
Type:
Antigen: BAFF-R
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human