Anti-FTO Mouse Monoclonal Antibody [clone: AG103]
Catalog # ADIPAG20A0091C050
Supplier: AdipoGen Life Sciences
Specifications
- Symbole de l'antigène:FTO
- Clonalité:Monoclonal
- Clone:AG103
- ELISA:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Isotype:IgG2a kappa
- Réactivité:Human
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:50 µG
Specifications
About this item
Type:
Antigen: FTO
Clonality: Monoclonal
Clone: AG103
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human