Anti-ANGPTL3 Rat Monoclonal Antibody [clone: Kairos3-1541]
N° de catalogue ADIPAG20A0087C100
Fournisseur: AdipoGen Life Sciences
Spécifications
- Symbole de l'antigène:ANGPTL3
- Clonalité:Monoclonal
- Clone:Kairos3-1541
- ELISA:Yes
- Hôte:Rat
- Isotype:IgG2a kappa
- Réactivité:Mouse
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:50 μg
- Cdt:100 µG
Spécifications
À propos de cet article
Type:
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: Kairos3-1541
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse