Anti-TNFRSF9 Rat Monoclonal Antibody [clone: M4173]
ADIPAG20A0071FC050
- Nom de l'antigène:Tumor Necrosis Factor Receptor Superfamily Member 9
- Symbole de l'antigène:TNFRSF9
- Clonalité:Monoclonal
- Clone:M4173
- Hôte:Rat
- Isotype:IgG1 kappa
- Réactivité:Mouse
- Western blot:Yes
- Formulaire:Liquid
- Synonymes antigène:CD137 antigen|CD137|T cell antigen ILA|T-cell antigen ILA|ILA|homolog of mouse 4-1BB|CDw137|4-1BB|T-cell antigen 4-1BB homolog|tumor necrosis factor receptor superfamily member 9|4-1BB ligand receptor
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:50 µG
Type:
Antigen: TNFRSF9
Clonality: Monoclonal
Clone: M4173
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse