Anti-IL17A Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 17-142F]
Catalog # ADIPAG20A0071C050
Supplier: AdipoGen Life Sciences
Specifications
- Nom de l'antigène:Interleukin 17A
- Symbole de l'antigène:IL17A
- Clonalité:Monoclonal
- Clone:17-142F
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Mouse
- Isotype:IgG1 kappa
- Réactivité:Human
- Formulaire:Liquid
- Synonymes antigène:IL-17|IL17|CTLA8|IL-17A|CTLA-8|Cytotoxic T-lymphocyte-associated serine esterase 8
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:50 μg
- Cdt:50 µG
Specifications
About this item
Type:
Antigen: IL17A
Clonality: Monoclonal
Clone: 17-142F
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human