Anti-ANGPTL4 Mouse Monoclonal Antibody [clone: Kairos4-397G]
Fournisseur: AdipoGen Life Sciences
Certificats
À propos de cet article
Type:
Antigen: ANGPTL4
Clonality: Monoclonal
Clone: Kairos4-397G
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Spécifications
- Catalog No:
- ADIPAG20A0046C050
- ADIPAG20A0046-C100
- Symbole de l'antigène:
- ANGPTL4
- ANGPTL4
- Clonalité:
- Monoclonal
- Monoclonal
- Clone:
- Kairos4-397G
- Kairos4-397G
- Réactivité:
- Human
- Human
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogène:
- Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Formulaire:
- Liquid
- Liquid
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- 0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- 0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:
- Short term: +4 °C, Long Term: –20 °C
- Short term: +4 °C, Long Term: –20 °C
Spécifications
Frequently Bought Together