Anti-ANGPTL6 Mouse Monoclonal Antibody [clone: Kairos-60]
N° de catalogue ADIPAG20A0039C100
Fournisseur: AdipoGen Life Sciences
Spécifications
- Symbole de l'antigène:ANGPTL6
- Clonalité:Monoclonal
- Clone:Kairos-60
- ELISA:Yes
- Hôte:Mouse
- Isotype:IgM kappa
- Réactivité:Human
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:0,5 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:50 μg
- Cdt:100 µG
Spécifications
À propos de cet article
Type:
Antigen: ANGPTL6
Clonality: Monoclonal
Clone: Kairos-60
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human