Anti-Listeria Mouse Monoclonal Antibody [clone: p6007]
Supplier: AdipoGen Life Sciences
Certificates
About this item
Type:
Antigen: Listeria
Clonality: Monoclonal
Clone: p6007
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Listeria monocytogenes
Specifications
- Catalog No:
- ADIPAG35B0006C100
- ADIPAG20A0022C050
- Symbole de l'antigène:
- Listeria
- Listeria
- Clonalité:
- Monoclonal
- Monoclonal
- Clone:
- p6007
- p6007
- Réactivité:
- Listeria monocytogenes
- Listeria monocytogenes
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogène:
- Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Formulaire:
- Liquid
- Liquid
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- 0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- 0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Température de stockage:
- Short term: +4 °C, Long Term: –20 °C
- Short term: +4 °C, Long Term: –20 °C
Specifications
Frequently Bought Together