Anti-VPS18 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1953R
Supplier: Bioss
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Vacuolar Protein Sorting 18
- Symbole de l'antigène:VPS18
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Adsorption croisée:No
- Formulaire:Liquid
- ID de gène:57617
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Température de stockage:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Température de transport:4 °C
- Immunogène:725-772/973
- Purification:Purified by Protein A
- Taille:100 μL
- Cdt:100 µl
Specifications
About this item
Type: Primary
Antigen: VPS18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat