ContactUsLinkComponent
Serum-free medium, MP-MRC-5®, MP Biomedicals
Serum-free medium, MP-MRC-5®, MP Biomedicals
Catalog # ICNA092013426
Supplier:  MP Biomedicals
CAS Number:  
Serum-free medium, MP-MRC-5®, MP Biomedicals
Catalog # ICNA092013426
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Format du milieu:
    Liquid
  • Cdt:
    1 L
  • Désignation:
    Serum-free medium, MP-MRC-5®
Specifications
About this item
MP-MRC-5® medium was developed specifically for the culture of MRC-5 cells and microcarrier culture of MRC-5 cells in production systems.
  • Serum-free, animal component free
  • Very low protein (<90 ng/ml)
  • Specific for MRC-5 cells