Human CD244 ELISA kit
Catalog # BIRBORB96808-96
Supplier: Biorbyt
CAS Number:
Specifications
- Cdt:96 Tests
- Primary antibody reactivity:Human
- Désignation:Human CD244 ELISA Kit
- Specificity:Natural and recombinant human CD244
- Portée de détection:62,5 - 4000 pg/ml
- Sensibilité:<10 pg/ml
Specifications
About this item
Sandwich ELISA kit for the quantitative detection of Human CD244 (Natural Killer Cell Receptor 2B4).
- No detectable cross-reactivity with other relevant proteins