Reference guide and photographs for steel surfaces, SSPC-VIS 4/NACE VIS 7
Codice catalogo SIMELD3058
Nome del fornitore: Industrial Physics
Specifiche
- Désignation:Guide and reference photographs for steel surfaces prepared by waterjetting
- Cdt:1
Specifiche
Informazioni su questo articolo
The SSPC-VIS 4/NACE VIS 7 shows six different steel surfaces - two uncoated and four previously coated - before and after waterjetting.
- Photographs illustrate four separate degrees of cleaning (WJ 1, WJ 2, WJ 3, and WJ 4) for each initial condition
- Additional photos that depict the appearance light, moderate, and heavy flash rust after cleaning
- Cleanliness requirements for conditions depicted are defined
- Joint SSPC/NACE surface preparation specification for high and ultra high pressure water jetting (SP 12/NACE 5)
Ordering information: Contains a written guide to the use of reference photographs and additional explanatory notes.