ContactUsLinkComponent
Reference guide and photographs for steel surfaces, SSPC-VIS 1
Reference guide and photographs for steel surfaces, SSPC-VIS 1
Catalog # SIMELD3055
Supplier:  Industrial Physics
undefined
Reference guide and photographs for steel surfaces, SSPC-VIS 1
Catalog # SIMELD3055
Supplier:  Industrial Physics
Specifications
  • Désignation:
    American photographic performance of the Swedish cleanliness by dry blasting
  • Cdt:
    1
Specifications
About this item
The SSPC is using visual reference, that features over 50 full colour pictures of (coated and uncoated) rusted steel surfaces, before and after dry abrasive blast cleaning.
Cleanliness requirements for conditions depicted are defined by the SSPC surface preparation specifications for white metal (SP 5), near-white (SP 10), commercial (SP 6), industrial (SP 14), and brush-off (SP 7) blast finishes. Appendix photographs show variations in white metal surfaces caused by different metallic and non-metallic abrasives, profile depth, angle of view, and lighting. Guides available in both English and Spanish.
Certifications: Similar to the Swedish and British standards, but the pictures of the required final appearances match the written descriptions of the US standards.