Reference manual for coating defects, Fitz's Atlas 2
Catalog # SIMELD3061
Supplier: Industrial Physics
Specifications
- Désignation:Fitz's Atlas 2 for coating defects, English only
- Cdt:1
Specifications
About this item
Fitz's Atlas 2 is a reference manual of colour images with a concise description of every problem and their associate causes.
The book is divided in sections such as weld faults, presurface conditions, coating defects, microscopy and marine fouling.