High visibility sleeve protectors, cut resistant, HyFlex® 11-202 HI-VIZ™
Catalog # ANSE11-202
Supplier: Ansell
Specifications
- Coupe du tenue:Regular
- Type:High visibility sleeve protectors
- Applications des tenues:Protection mécanique
- Genre:Unisex
- Zone à risque:Résistant aux coupures,Haute visibilité
- Couleur:jaune hi-viz
- Longueur:475 mm
- Tissu:polyester, nylon and glass
- Cdt:12
Specifications
About this item
This sleeve made of polyester, nylon and glass fibre has a level EN CUT 5 performance and an increased abrasion protection. Safety is also enhanced through its visibility colour and level 1 thermal protection.
- Sleeve delivers the best comfort through its anatomical design
- Offers an improved fit thanks to its adjustable VELCRO® strap system
- Freedom of movement leads to increased productivity and worker acceptance
Ideal for use in body shops (excluding welding), for glass and metal sheet handling and for stamping.
Gauge: 13
Certifications: EN 388 (2542), EN 407 (X1XXXX); Cat. III