Test kits, water balance, Comparator 2000+
Test kits, water balance, Comparator 2000+
Water balance test kit, 0,2 - 4,0 mg/l (Cl2); 6,8 - 8,4 pH; 10 - 500 mg/l (total alkalinity); 10 - 500 mg/l CaCO3 (calcium hardness), AF 129, 1 KIT
N° de catalogue LOVI411290
Fournisseur:   Lovibond
Test kits, water balance, Comparator 2000+
N° de catalogue LOVI411290
Fournisseur:  Lovibond
Numéro CAS:  
Test kits, water balance, Comparator 2000+
Test kits, water balance, Comparator 2000+
Water balance test kit, 0,2 - 4,0 mg/l (Cl2); 6,8 - 8,4 pH; 10 - 500 mg/l (total alkalinity); 10 - 500 mg/l CaCO3 (calcium hardness), AF 129, 1 KIT
Numéro CAS:  
Prix
SFr. 1’490.00
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Informations de livraison: Kit includes Comparator 2000+, pH comparator disc, chlorine comparator disc, 4× Comparator vials (10 ml), 13 mm path length shaker tubes, tablet reagents for each parmeter, crushing / stirring rod, vial cleaning brush, 100 ml graduated cylinder, instruction sheet and Langelier index / balanced water calculation table.
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    Water balance test kit
  • Type:
    AF 129
  • Plage de mesure:
    0,2 - 4,0 mg/l (Cl<sub>2</sub>); 6,8 - 8,4 pH; 10 - 500 mg/l (total alkalinity); 10 - 500 mg/l CaCO<sub>3 </sub>(calcium hardness)
Spécifications