Test kits, amine, Comparator 2000+
N° de catalogue LOVI412740
Fournisseur: Lovibond
Numéro CAS:
Spécifications
- Cdt:1 KIT
- Désignation:Amines test kit
- Seq:01
- Type:AF 274
- Plage de mesure:1 - 10 mg/l
Spécifications
À propos de cet article
Comparator 2000+ is a versatile modular system for testing water.
- Stable and accurate
- Robust, compact and portable
- Prism enabling colour matching
- Diffuser plate is used to measure the turbidity
- Longer path length cells be used for lower concentration analysis
Livraison: Test kit is supplied in a robust carrying case containing Comparator 2000+, amines comparator disc, extraction tube for result measurement, filter funnel and filter papers, 100 ml amber glass bottle, tablet reagents, crushing/stirring rod and instruction sheet.