ContactUsLinkComponent
Solution for junction cleaning, KS410 (Radiometer Analytical)
Catalog # HACHC20C380
Supplier:  Hach
undefined
Solution for junction cleaning, KS410 (Radiometer Analytical)
Catalog # HACHC20C380
Supplier:  Hach
Specifications
  • Désignation:
    Thiourea solution for junction cleaning, for combined/reference electrodes, KS410, 250 ml bottle
  • Seq:
    03
  • Cdt:
    250 mL