Sealed cell assemblies
Permanently sealed omni-cell window units, CaF2, 0,20 mm thickness
N° de catalogue SPCC1942
Fournisseur:   Specac
Sealed cell assemblies
N° de catalogue SPCC1942
Fournisseur:  Specac
Sealed cell assemblies
Permanently sealed omni-cell window units, CaF2, 0,20 mm thickness
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 Un.
Product Details & Documents
Rectangular with lead spacer sealed cell assemblies for omni cell units.
Spécifications
  • Désignation:
    Permanently sealed omni-cell window units, CaF2, 0,20 mm thickness
  • Seq:
    09