ContactUsLinkComponent
Buffer solution for water testing
Catalog # HACH1430598
Supplier:  Hach
CAS Number:  
undefined
Buffer solution for water testing
Catalog # HACH1430598
Supplier:  Hach
CAS Number:  
Specifications
  • Cdt:
    25
  • Désignation:
    Magnesium chloride/potassium phosphate buffer solution, 99 ml
Specifications
About this item
Buffered dilution water required for testing recreational, drinking and wastewater.