Cation exchange HPLC columns, Dionex™, IonPac™ CS17
Fournisseur: DIONEX
Certificats
À propos de cet article
The Dionex™, IonPac™ CS17 column is a universal column for the separation of polyvalent and hydrophobic amines in complex sample matrices. It is mainly used for analysis of power plant waters treated with ammonium, morpholine, or ethanolamine, chemical additives, chemical process solutions, scrubber solutions, plating baths, industrial solvents, and food samples.
Frequently Bought Together