ContactUsLinkComponent
Condensers, Allihn, with Quickfit® ground joints
  201-8312
undefined
Condensers, Allihn, with Quickfit® ground joints
  201-8312
  • Rodage mâle:
    24/29* NS
  • Longueur du condenseur:
    400 mm
  • Rodage femelle:
    24/29 NS
  • Seq:
    07
  • Cdt:
    1
Condensers acc. to Allihn, PYREX® borosilicate glass, for condensing vapours.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for reflux applications. With NS socket and NS cone. Inner bulb sections provide increased cooling surface. With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing.