Laboratory glass cleaner, deconex®16 PLUS
DECO509700.00-K05W
- Désignation:deconex® 16 PLUS
- Seq:31
- Cdt:5 kg
Liquid, pH-neutral cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials for use in ultrasonic and soaking baths.
- Rinsable without residues and guarantees therefore consistent and error-free results
- Perfect results even at low concentrations
- Working solution suitable for multiple use
- Good material compatibility, especially recommended for alkali and corrosion sensitive laboratory material, also suitable for aluminium
- Phosphate-free, no release of phosphates into waste water
- EDTA-, NTA- and chlorine-free, environmentally friendly