Dust covers
Dust covers, for VOA vial caps, 24 - 414 mm
N° de catalogue EPSCDC-VOA
Fournisseur: EP SCIENTIFIC
Aller à
Product Details & Documents
These PE dust covers are designed to fit all standard size VOA vial caps. They prevent contamination and protect critical samples during transport.
- Allow for septum expansion
- Pre cleaned dust covers available upon request
Spécifications
- Taille du bouchon:24 - 414 mm
- Désignation:Dust covers, for VOA vial caps
- Cat. No.:EPSCDC-VOA
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact