ContactUsLinkComponent
Biotin SE protein labelling kit
Biotin SE protein labelling kit
Catalog # 92224.
Supplier:  Biotium
CAS Number:  
Biotin SE protein labelling kit
Catalog # 92224.
Supplier:  Biotium
CAS Number:  
Specifications
  • Cdt:
    1 KIT
  • Désignation:
    Biotin SE Protein Labeling Kit, for three labeling reactions, up to 1 mg protein per reaction
Specifications
About this item
This kit contains everything needed to label antibodies or other proteins with biotin succinimidyl ester (biotin SE). Each kit contains reagents for three labelling reactions of up to 1 mg protein per reaction.