ContactUsLinkComponent
VISOCOLOR® Reagent Case with Accessories for Soul Analysis
VISOCOLOR® Reagent Case with Accessories for Soul Analysis
  MANA934220
 :  
undefined
VISOCOLOR® Reagent Case with Accessories for Soul Analysis
  MANA934220
 :  
  • Cdt:
    1
  • Désignation:
    VISOCOLOR® Reagent case for soil analysis with PF-3
  • Dimensions:
    525×455×200 mm (including packaging)
  • Seq:
    0
  • Température de stockage:
    15…25 °C
  • Durée de conservation (mois):
    18
  • Poids:
    8900 g (including packaging)
  • Type:
    Hazardous material: Yes
  • Tests par set:
    1 pieces
  • Plage de mesure:
    0
The VISOCOLOR® soil case with PF-3 soil is the perfect tool for reliable and simple determination of soil samples.
  • Combines all important measurements in one handy device
  • Equipped with the necessary accessories for sample preparation of soil samples and reagents
  • For the determination of ammonium, potassium, nitrate, phosphate and pH
Equipped with a filter photometer, the analysis case enables both visual and photometric evaluation.