ContactUsLinkComponent
Immobilised TCEP disulphide reducing gel, Pierce™
Immobilised TCEP disulphide reducing gel, Pierce™
N° de catalogue PIER77712
Numéro CAS:  
undefined
Immobilised TCEP disulphide reducing gel, Pierce™
N° de catalogue PIER77712
Numéro CAS:  
Spécifications
  • Cdt:
    5 ml
  • Désignation:
    Immobilised TCEP disulfide reducing gel
Spécifications
À propos de cet article
Immobilised TCEP disulphide Reducing Gel eliminates the need to use laborious and inefficient gel filtration methods to separate the reduced sample from the reducing agent.
  • Odourless and virtually no loss of activity when stored refrigerated
  • Reduces peptides and proteins over a broad range of pH, salt or detergent concentrations, and temperature conditions
  • Resin slurry provides flexibility for use in spin tube, column or batch modes
  • No soluble reductant removal saves time and conserves sample
  • Cleaves both natural disulfide bonds and those incorporated by crosslinking