Affinity purification kits, Melon™ Gel spin plate kit for IgG screening
Catalog # PIER45208
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Cdt:2
- Désignation:Melon™ Gel Spin Plate Kit for IgG Screening, sufficient to process up to 192 samples containing 20 - 100 µl
Specifications
About this item
The Melon™ Gel Spin Plate Kit for IgG Screening offers a high throughput format for quick purification and screening of antibodies.
- Simple, one-step protocol with rapid purification
- Minimal dilution, reduced need to concentrate antibody before storage or use
- Gentle purification, no harsh antibody elution conditions
- Melon™ gel resin can be used for multiple antibody purifications
Melon™ Gel IgG Purification Kits enable purification of antibodies from serum and other sample types. Melon™ Gel Resin binds and removes serum proteins, thereby allowing pure IgG to be collected in the flow-through fraction.
Delivery: Melon™ gel 96-well spin plates, 2 each, containing 100 µl of Melon™ gel/well; Melon gel purification buffer, 2×100 ml; wash plates (white), 2 each; and collection plate.