ContactUsLinkComponent
Affinity purification resins, UltraLink™ iodoacetyl resin
Affinity purification resins, UltraLink™ iodoacetyl resin
Catalog # PIER53155
CAS Number:  
undefined
Affinity purification resins, UltraLink™ iodoacetyl resin
Catalog # PIER53155
CAS Number:  
Specifications
  • Cdt:
    10 mL
  • Désignation:
    UltraLink™ Iodoacetyl Resin
  • Type:
    Liquid
Specifications
About this item
UltraLink™ iodoacetyl resin is a durable, porous resin that is activated to enable efficient and direct covalent immoblisation of peptides and other ligands through their sulfhydryl groups (-SH) for use in affinity purification procedures.
  • High coupling efficiency and capacity
  • Specific and leak-proof coupling chemistry
  • Durable, high-performance resin
  • Prepared affinity resin can be used with typical binding and elution procedures for many cycles of affnity purification
The bead structure and efficiently coupling chemistry of the iodoacetyl-activated UltraLink™ support results in high protein binding capacity, high linear flow rates, low non specific binding and overall superior performance in affinity chromatography. UltraLink™ resins are ideal for medium pressure applications, such as FPLC.