ContactUsLinkComponent
HA-Tag IP/Co-IP kit, Pierce™
HA-Tag IP/Co-IP kit, Pierce™
Catalog # PIER26180
CAS Number:  
undefined
HA-Tag IP/Co-IP kit, Pierce™
Catalog # PIER26180
CAS Number:  
Specifications
  • Cdt:
    1 KIT
  • Désignation:
    Pierce™ HA Tag IP/Co-IP Kit, sufficient for 25 IP/Co-IP reactions each using 10 µl of resin
Specifications
About this item
Pierce™ HA-Tag IP/Co-IP kit provides the affinity resin, positive control and other reagents necessary to perform immunoprecipitation (IP) or co-immunoprecipitation (Co-IP) reactions with an HA-tagged bait protein.
  • Highly specific anti-HA monoclonal antibody provides high-yield immunoprecipitation products
  • The kit and anti-HA agarose are compatible with various cell lysates and physiologic buffer systems
This kit is based on crosslinked beaded agarose to which a highly specific anti-HA antibody is covalently immobilised. The ready-to-use affinity resin, together with the included buffers, microcentrifuge spin columns, positive control, and easy-to-follow instructions, constitute a complete set of reagents sufficient for 25 for IP or Co-IP assays.
Delivery: Kit includes Anti-HA agarose, 0,25 ml; HA-tagged positive control, 0,5 ml; BupH Tris buffered saline pack, 500 ml; Lane Marker Non-reducing Sample Buffer (5X), 5 ml; spin columns accessory pack, 27 columns; and collection tubes and caps accessory pack, 100 tubes.