Ultrapure lyophilisation stopper, WHEATON®
Stopper with 3 legs, 20,0 mm
N° de catalogue WHEAW224100-407
Fournisseur: DWK Life Sciences
Ultrapure lyophilisation stopper, WHEATON®
N° de catalogue WHEAW224100-407
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
This ultrapure lyophilization stopper is a very low moisture adsortion stopper. Manufactured from an ultra pure bromobutyl formulation with extremely high chemical purity and low gas permeability.
- Very low level of adsortion of the preservatives present in aqueous parenteral solutions
- Autoclavable
- Free of latex
Spécifications
- Ø inférieur:20,0 mm
- Désignation:Stopper with 3 legs