Hardness reagent, UniVer® 3
Catalog # HACH21320H
Supplier: Hach
CAS Number:
Specifications
- Méthode chimique:EDTA
- Cdt:28,3 g
- Désignation:Hardness reagent, UniVer® 3
Specifications
About this item
This combined indicator and buffer reagent is used for determination of total hardness by titration methods.