ContactUsLinkComponent
Tablet count kits for water hardness
Tablet count kits for water hardness
Catalog # PALIAD-1016
Supplier:  Palintest
CAS Number:  
undefined
Tablet count kits for water hardness
Catalog # PALIAD-1016
Supplier:  Palintest
CAS Number:  
Specifications
  • Cdt:
    250
  • Désignation:
    Total hardness test, 250 tests
Specifications
About this item
Tablet Count kits provide a simple method of analysis for an important range of water parameters. The tests are particularly useful for the routine control of industrial waters such as boiler waters and cooling water systems, as well as swimming pools, natural waters and similar applications.
  • Cost effective and provides quick results on-site
  • Can be used as a stand alone kit or combined with comparator or photometer
  • Wide range of test concentrations can be measured