Test Kits, oxygen scavengers, model OS-1
Catalog # HACH2349500
Supplier: Hach
CAS Number:
Specifications
- Méthode chimique:Iron reduction
- Cdt:50 Tests
- Désignation:Oxygen scavengers test kit OS-1
- Pour appareil:hydroquinone, ISA, MEKO
Specifications
About this item
OS-1 Kit provides proper maintenance and economy of operation of modern boilers and cooling towers which requires balance between water quality parameters and treatment chemicals. It measures common corrosion and biological growth inhibitor using simple color disc method.
Delivery: Kit includes color disc, comparator box and tubes, reagents, mixing bottles, instructions and carrying case.