ContactUsLinkComponent
Kits de tests, molybdène Spectroquant®, Supelco®
Kits de tests, molybdène Spectroquant®, Supelco®
Catalog # 1.00860.0001
Supplier:  Merck
CAS Number:  
undefined
Kits de tests, molybdène Spectroquant®, Supelco®
Catalog # 1.00860.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Méthode chimique:
    Rouge de bromopyrogallol
  • Cdt:
    25 Tests
  • Désignation:
    Molybdène, tubes
  • Seq:
    01
  • M-Clarity quality level:
    MQ100
  • Pour appareil:
    Prove, Nova 30/60, Multy
  • Plage de mesure:
    0,02 - 1,00 mg/l Mo
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Kits de test de réactifs Spectroquant®.
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.