Software for data loggers, elproLOG ANALYZE
Software for data loggers, elproLOG ANALYZE
Supplier:   ELPRO-BUCHS
Software for data loggers, elproLOG ANALYZE
Supplier:  ELPRO-BUCHS
Certificates
Software for data loggers, elproLOG ANALYZE
2 Options Available Below
Product Details & Documents
elproLOG ANALYZE is a software for all elpro data loggers. It provides all functions such as data analysis, reporting and configuration of data loggers.
  • User friendly
  • Displays statistics on measurement data
  • Optional audit trail
  • Multiple zoom function
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • elproLOG ANALYZE software, full version
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • elproLOG ANALYZE software, update