Safety ankle boots, lace-up, Concept Plus, Jazz
Catalog # UPOWUC10094/43
Supplier: U-Power
Specifications
- Pointure:43
- Couleur:ambré
- Cdt:1 PAIR
Specifications
About this item
Metal-free, anti-static safety ankle boots made from pull-up water repellent leather and a hard wearing, slip resistant PU sole.
- Action dry light, non slip insole made from EVA/textile keeps feet dry; anti-fungal treated
- WingTex tunnel lining, with honeycomb structure, ensures breathability and excellent absorbent features
- Stress-out tongue system allows easy instep providing added comfort when walking
- Natural fit (width 11) for additional comfort
- AirToe protective aluminium toecap with breathable membrane (200 J)
- Light, flexible and non metallic, puncture resistant midsole Save & Flex covers 100% of the sole
Colour: Brown
Colour: Brown
Certifications: EN 20345:2011; S3, SRC.