Racks and boxes
 :   TPP
Racks and boxes
 :  TPP
Racks and boxes
Racks and boxes
These reusable cryotube racks and boxes with alphanumerical inscription are ideal for sample identification.
  • Polypropylene, heat resistant, autoclavable material
  • Racks can be quickly assembled and disassembled thus space saver
  • Cryo-box, single handling of lid even after multiple use
  • Cryo-rack, locking system in the groves allowing one hand operation when used with star-shape foot cryo tube
 : ISO 9001:2008 certified.
Product Family Options
  • Désignation
    Pour tubes
    Nb de places
    Couleur
    l×p×h
  • Box for cryotubes
    1,2/2 ml
    81
    jaune
    133×133×45 mm
    1 * 1 ST
    1 * 1 ST - SFr. 29.40
    SFr. 29.40
    1 * 1 ST
       SFr. 29.40
  • Box for cryotubes
    4/5 ml
    81
    jaune
    133×133×95 mm
    1 * 1 ST
    1 * 1 ST - SFr. 30.70
    SFr. 30.70
    1 * 1 ST
       SFr. 30.70
  • Portoir pour tubes de culture tissulaire
    5 ml
    8
    jaune
    200×88×72 mm
    1 * 1 ST
    1 * 1 ST - SFr. 10.50
    SFr. 10.50
    1 * 1 ST
       SFr. 10.50
  • Rack for cryotubes
    15/50 ml
    18/10
    jaune
    209×98×62 mm
    1 * 1 ST
    1 * 1 ST - SFr. 10.50
    SFr. 10.50
    1 * 1 ST
       SFr. 10.50
  • Rack for cryotubes with star foot
    40
    jaune
    219×109×26 mm
    1 * 1 ST
    1 * 1 ST - SFr. 29.40
    SFr. 29.40
    1 * 1 ST
       SFr. 29.40
  • Rack "L" for centrifuge tubes
    15/50 ml
    30/20
    jaune
    209×172×62 mm
    1 * 1 ST
    1 * 1 ST - SFr. 10.50
    SFr. 10.50
    1 * 1 ST
       SFr. 10.50
  • Rack for cryotubes
    600 ml
    3
    jaune
    366×146×71 mm
    1 * 6 ST
    1 * 6 ST - SFr. 174.00
    SFr. 174.00
    1 * 6 ST
       SFr. 174.00